Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MFGACYKQPL KPSGSEPPAE ECRMTPRHAG CDVTEMQRIL SQPTFTEHLL RAV
HHLA3 (HERV-H LTR-Associating 3) is an RNA Gene, and is affiliated with the lncRNA class. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: human endogenous retrovirus-H long terminal repeat-associating 3; human endogenous retrovirus-H long terminal repeat-associating protein 3
Entrez Gene ID: (Human) 11147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support