Oxoid™ Microbact™ GNB Reagents and Accessories
Actual product may vary
Thermo Scientific™

Oxoid™ Microbact™ GNB Reagents and Accessories

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
MB0184AMicrobact Reagent VP II10 mL/Bottle
MB0266AMicrobact Oxidase Strip50/Pk.
MB5370AMicrobact Holding Tray 12A/12B1 Tray
MB0181AMicrobact Reagent VP I10 mL/Bottle
MB0180AMicrobact Reagent TDA10 mL/Bottle
MB1082AMicrobact Reagent Set D6 x 10 mL Bottles
MB0187AMicrobact Reagent Nitrate B10 mL/Bottle
MB5128AMicrobact Holding Tray 24E/12E1 Tray
MB1448AMicrobact Reagent Spot Indole DMACA10 mL/Bottle
MB1093AMicrobact Mineral Oil1 Bottle
MB0209AMicrobact Reagent Indole-Kovacs10 mL/Bottle
MB0186AMicrobact Reagent Nitrate A10 mL/Bottle
Catalog number MB0184A
Price (MXN)
-
Description:
Microbact Reagent VP II
Quantity:
10 mL/Bottle
Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.
Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

General References:

  1. Farmer, J.J. et al. (1985) J. Clin. Micro. 21(1): 46-76
  2. Burke, V., Robinson, J., Atkinson, H.M. and Gracey, M. (Jan 1982) J. Clin. Micro. 48-52
  3. Cowen, S.T. and Steel, K.J. (1977) Manual for the Identification of Medical Bacteria, 2nd edition, Cambridge University Press.
  4. Balows, A., Hausler, W.J., Herrmann, K.L., Isengerg, J.D. and Shadomy, H.J. (eds) (1991) Manual of Clinical Microbiology, 5th edition, American Society of Microbiology, Washington
Specifications
DescriptionMicrobact Reagent VP II
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach