Thermo Scientific™

Oxoid™ Microbact™ GNB Reagents and Accessories

Catalog number: MB0187A
Have Questions?
Thermo Scientific™

Oxoid™ Microbact™ GNB Reagents and Accessories

Catalog number: MB0187A
Have Questions?
Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.
 
Catalog Number
MB0187A
Unit Size
Each
Description
Microbact Reagent Nitrate B
Quantity
10 mL/Bottle
Price (USD)
Full specifications
DescriptionMicrobact Reagent Nitrate B
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Reagent VP II
Quantity
10 mL/Bottle
Price (USD)
Full specifications
DescriptionMicrobact Reagent VP II
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Oxidase Strip
Quantity
50/Pk.
Price (USD)
Full specifications
DescriptionMicrobact Oxidase Strip
FormatMicroplate, Strip
Packaging Type1 Box
Quantity50/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Holding Tray 12A/12B
Quantity
1 Tray
Price (USD)
Full specifications
DescriptionMicrobact Holding Tray 12A/12B
FormatMicroplate, Strip
Packaging TypeEach
Quantity1 Tray
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Reagent VP I
Quantity
10 mL/Bottle
Price (USD)
Full specifications
DescriptionMicrobact Reagent VP I
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Reagent TDA
Quantity
10 mL/Bottle
Price (USD)
Full specifications
DescriptionMicrobact Reagent TDA
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Reagent Set D
Quantity
6 x 10 mL Bottles
Price (USD)
Full specifications
DescriptionMicrobact Reagent Set D
FormatBottle
IncludesMicrobact Reagent TDA, VP I, VP II; Nitrate A; Nitrate B; Indole-Kovacs
Packaging Type1 Box
Quantity6 x 10 mL Bottles
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Holding Tray 24E/12E
Quantity
1 Tray
Price (USD)
Full specifications
DescriptionMicrobact Holding Tray 24E/12E
Detectable AnalytesListeria species
FormatMicroplate, Strip
Packaging TypeEach
Quantity1 Tray
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Reagent Spot Indole DMACA
Quantity
10 mL/Bottle
Price (USD)
Full specifications
DescriptionMicrobact Reagent Spot Indole DMACA
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Microbact Mineral Oil
Quantity
1 Bottle
Price (USD)
Full specifications
DescriptionMicrobact Mineral Oil
FormatBottle
Packaging Type1 Box
Quantity1 Bottle
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)
MB0187AFull specifications
EachMicrobact Reagent Nitrate B10 mL/BottleRequest A Quote
DescriptionMicrobact Reagent Nitrate B
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
MB0184AFull specifications
EachMicrobact Reagent VP II10 mL/BottleRequest A Quote
DescriptionMicrobact Reagent VP II
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
MB0266AFull specifications
EachMicrobact Oxidase Strip50/Pk.Request A Quote
DescriptionMicrobact Oxidase Strip
FormatMicroplate, Strip
Packaging Type1 Box
Quantity50/Pk.
Unit SizeEach
MB5370AFull specifications
EachMicrobact Holding Tray 12A/12B1 TrayRequest A Quote
DescriptionMicrobact Holding Tray 12A/12B
FormatMicroplate, Strip
Packaging TypeEach
Quantity1 Tray
Unit SizeEach
MB0181AFull specifications
EachMicrobact Reagent VP I10 mL/BottleRequest A Quote
DescriptionMicrobact Reagent VP I
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
MB0180AFull specifications
EachMicrobact Reagent TDA10 mL/BottleRequest A Quote
DescriptionMicrobact Reagent TDA
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
MB1082AFull specifications
EachMicrobact Reagent Set D6 x 10 mL BottlesRequest A Quote
DescriptionMicrobact Reagent Set D
FormatBottle
IncludesMicrobact Reagent TDA, VP I, VP II; Nitrate A; Nitrate B; Indole-Kovacs
Packaging Type1 Box
Quantity6 x 10 mL Bottles
Unit SizeEach
MB5128AFull specifications
EachMicrobact Holding Tray 24E/12E1 TrayRequest A Quote
DescriptionMicrobact Holding Tray 24E/12E
Detectable AnalytesListeria species
FormatMicroplate, Strip
Packaging TypeEach
Quantity1 Tray
Unit SizeEach
MB1448AFull specifications
EachMicrobact Reagent Spot Indole DMACA10 mL/BottleRequest A Quote
DescriptionMicrobact Reagent Spot Indole DMACA
FormatBottle
Packaging Type1 Box
Quantity10 mL/Bottle
Unit SizeEach
MB1093AFull specifications
EachMicrobact Mineral Oil1 BottleRequest A Quote
DescriptionMicrobact Mineral Oil
FormatBottle
Packaging Type1 Box
Quantity1 Bottle
Unit SizeEach
Showing 10 of 12Show more
Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
1. Farmer, J.J. et al. (1985) J. Clin. Micro. 21(1): 46-76
2. Burke, V., Robinson, J., Atkinson, H.M. and Gracey, M. (Jan 1982) J. Clin. Micro. 48-52
3. Cowen, S.T. and Steel, K.J. (1977) Manual for the Identification of Medical Bacteria, 2nd edition, Cambridge University Press.
4. Balows, A., Hausler, W.J., Herrmann, K.L., Isengerg, J.D. and Shadomy, H.J. (eds) (1991) Manual of Clinical Microbiology, 5th edition, American Society of Microbiology, Washington

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text