Search
Search
| Número de catálogo | Longitud de onda |
|---|---|
| MEOF0660 | 660 nm |
| MEOF0414 | 414 nm |
| MEOF0492 | 492 nm |
| MEOF0520 | 520 nm |
| MEOF0540 | 540 nm |
| MEOF0570 | 570 nm |
| MEOF0600 | 600 nm |
| MEOF0690 | 690 nm |
| MEOF0750 | 750nm |
The Thermo Scientific™ filter accessories for the Multiskan™ Ease Absorbance Microplate Reader expand assay flexibility by enabling precise wavelength selection across a wide range of absorbance applications. Designed to integrate smoothly with the instrument's optical system, these durable hard-coated filters support both routine and advanced assays while maintaining high measurement accuracy and long-term reliability.
The Multiskan™ Ease utilizes an easy-to-install 8-position filter wheel, supplied with three pre-installed standard filters (405 nm, 450 nm, and 620 nm). Additional filter accessories allow users to customize wavelength configurations to meet specific assay requirements, from ELISA and protein quantification to enzyme kinetics and microbial growth analysis.
Optical Filter Wavelengths and Typical Applications
Standard (pre-installed on filter wheel):
Optional filters:
Key Features
These optical filters and filter wheel are designed for use with the Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader. They support assay-specific wavelength optimization to enhance data quality and are fully integrated with Thermo Scientific™ SkanIt™ Software for streamlined assay setup and data analysis within existing laboratory workflows.