EpiLife™ Medium, with 60 μM calcium
Gibco™
EpiLife™ Medium, with 60 μM calcium
EpiLife is a sterile, liquid medium prepared with 60 μM calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells.
Have Questions?
Catalog NumberQuantity
MEPI500CA500 mL
Catalog number MEPI500CA
Price (USD)
129.65
Online exclusive
139.00
Save 9.35
Each
Quantity:
500 mL
Price (USD)
129.65
Online exclusive
139.00
Save 9.35
Each
EpiLife is a sterile, liquid medium prepared with 60 μM calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of an appropriate growth supplement prior to use. EpiLife Medium greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin-free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife Medium is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.
To support plating and long term proliferation of human keratinocytes, EpiLife Medium must be supplemented with selected hormones and growth factors. We offer a choice of growth supplements for culturing human keratinocytes. Please refer to the EpiLife Medium user guide to determine the best supplement choice for your purposes. The supplements contain all of the growth factors, hormones, and extracts necessary for growth of human keratinocytes in EpiLife Medium.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeKeratinocyte Cells, Epithelial Cells
Inorganic SaltsCalcium
Product LineCascade Biologics, EpiLife
Product TypeEpiLife Medium
Quantity500 mL
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free, cGMP
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHEPES, Phenol Red
Without AdditivesNo Antibiotics
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Frequently asked questions (FAQs)
Are the Keratinocyte SFM basal medium and supplement kit available separately?
The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture mouse keratinocytes?
The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture human keratinocytes?
There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.