Thermo Scientific™
Meridian™ 7 System for Semiconductors
Catalog number: MERIDIAN7
The Thermo Scientific™ Meridian™ 7 system provides visible laser voltage imaging and probing and dynamic laser stimulation on sub-10nm devices. By avoiding a requirement for ultra-thin substrates, it preserves the integrity and functionality of the device under test to provide a reliable and practical production solution. It offers a 25 percent optical resolution enhancement over the previous-generation system and has a smaller spot size for better fault localization. In addition, the new Meridian 7 system offers more certainty in navigation and computer-aided design (CAD) overlay, less cross-talk and higher waveform signal-to-noise.