Meridian™ V System for Semiconductors
Meridian™ V System for Semiconductors
Meridian™ V System for Semiconductors
Thermo Scientific™
Meridian™ V System for Semiconductors
Optimized dynamic LSM platform
Have Questions?
Número de catálogo MERIDIANV
Precio (MXN)
-
Optimized dynamic LSM platform
  • Performance optical path for best imaging
  • Architecture supports automated test equipment (ATE) docking and back-side (B/S) analysis
  • Mobility for the lab; high power cooling options
Best-in-class performance
  • Laser voltage probing (LVP) and laser voltage imaging (LVI) for timing and functional analysis
  • Dynamic laser simulation (DLS) for critical path analysis
  • Powerful and easy-to-use Sierra Software GUI
Dynamic electrical fault isolation
  • Transistor- and cell-level fault localization
  • Isolate design-process marginalities
  • Based on Ruby optics
Configuration & Features
Highest performance solid immersion lens (SIL) and imaging
  • Optimized monochromatic optical path
  • 'Point & Click' high numerical aperture (NA) SIL (3.1, 2.9NA)
  • Polarization-optimized imaging
  • High performance 1320nm LVI and LVP
  • Low frequency and high bandwidth options
  • High resolution 1064nm LVx option
Performance confocal LSM
  • Static and dynamic laser stimulation; SLS Lock-in
  • Dual wavelength near infrared (NIR system) (1064, 1320/1340)
System
  • Sierra software suite
  • Based on Ruby optics
Especificaciones
Product Size-
Unit SizeEach