Product References
An instantly fixable and self-adaptive scaffold for skull regeneration by autologous stem cell recruitment and angiogenesis.
Nature communications
Lu G,Xu Y,Liu Q,Chen M,Sun H,Wang P,Li X,Wang Y,Li X,Hui X,Luo E,Liu J,Jiang Q,Liang J,Fan Y,Sun Y,Zhang X
MF48005 was used in Flow Cytometry to design an instantly fixable and self-adaptive scaffold by dopamine-modified hyaluronic acid chelating Ca2+ of the microhydroxyapatite surface and bonding type I collagen to highly simulate the natural bony matrix.
Fri May 06 00:00:00 EDT 2022
Tick extracellular vesicles enable arthropod feeding and promote distinct outcomes of bacterial infection.
Nature communications
Oliva Chávez AS,Wang X,Marnin L,Archer NK,Hammond HL,Carroll EEM,Shaw DK,Tully BG,Buskirk AD,Ford SL,Butler LR,Shahi P,Morozova K,Clement CC,Lawres L,Neal AJO,Mamoun CB,Mason KL,Hobbs BE,Scoles GA,Barry EM,Sonenshine DE,Pal U,Valenzuela JG,Sztein MB,Pasetti MF,Levin ML,Kotsyfakis M,Jay SM,Huntley JF,Miller LS,Santambrogio L,Pedra JHF
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Thu Jun 17 00:00:00 EDT 2021
Tick extracellular vesicles enable arthropod feeding and promote distinct outcomes of bacterial infection.
Nature communications
Oliva Chávez AS,Wang X,Marnin L,Archer NK,Hammond HL,Carroll EEM,Shaw DK,Tully BG,Buskirk AD,Ford SL,Butler LR,Shahi P,Morozova K,Clement CC,Lawres L,Neal AJO,Mamoun CB,Mason KL,Hobbs BE,Scoles GA,Barry EM,Sonenshine DE,Pal U,Valenzuela JG,Sztein MB,Pasetti MF,Levin ML,Kotsyfakis M,Jay SM,Huntley JF,Miller LS,Santambrogio L,Pedra JHF
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Thu Jun 17 00:00:00 EDT 2021
Tumor-Derived Retinoic Acid Regulates Intratumoral Monocyte Differentiation to Promote Immune Suppression.
Cell
Devalaraja S,To TKJ,Folkert IW,Natesan R,Alam MZ,Li M,Tada Y,Budagyan K,Dang MT,Zhai L,Lobel GP,Ciotti GE,Eisinger-Mathason TSK,Asangani IA,Weber K,Simon MC,Haldar M
MF48005 was used in Flow cytometry/Cell sorting to find that the TME induces tumor cells to produce retinoic acid (RA), which polarizes intratumoral monocyte differentiation toward TAMs and away from DCs via suppression of DC-promoting transcription factor Irf4.
Thu Mar 19 00:00:00 EDT 2020
Nanoparticle Vaccines Based on the Receptor Binding Domain (RBD) and Heptad Repeat (HR) of SARS-CoV-2 Elicit Robust Protective Immune Responses.
Immunity
Ma X,Zou F,Yu F,Li R,Yuan Y,Zhang Y,Zhang X,Deng J,Chen T,Song Z,Qiao Y,Zhan Y,Liu J,Zhang J,Zhang X,Peng Z,Li Y,Lin Y,Liang L,Wang G,Chen Y,Chen Q,Pan T,He X,Zhang H
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Tue Dec 15 00:00:00 EST 2020
Nanoparticle Vaccines Based on the Receptor Binding Domain (RBD) and Heptad Repeat (HR) of SARS-CoV-2 Elicit Robust Protective Immune Responses.
Immunity
Ma X,Zou F,Yu F,Li R,Yuan Y,Zhang Y,Zhang X,Deng J,Chen T,Song Z,Qiao Y,Zhan Y,Liu J,Zhang J,Zhang X,Peng Z,Li Y,Lin Y,Liang L,Wang G,Chen Y,Chen Q,Pan T,He X,Zhang H
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Tue Dec 15 00:00:00 EST 2020
Intratumoral Monocyte Transfer to Examine Monocyte Differentiation in the Tumor Microenvironment.
STAR protocols
Devalaraja S,Haldar M
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Fri Dec 18 00:00:00 EST 2020
Intratumoral Monocyte Transfer to Examine Monocyte Differentiation in the Tumor Microenvironment.
STAR protocols
Devalaraja S,Haldar M
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Fri Dec 18 00:00:00 EST 2020
Counter Regulation of Spic by NF-κB and STAT Signaling Controls Inflammation and Iron Metabolism in Macrophages.
Cell reports
Alam Z,Devalaraja S,Li M,To TKJ,Folkert IW,Mitchell-Velasquez E,Dang MT,Young P,Wilbur CJ,Silverman MA,Li X,Chen YH,Hernandez PT,Bhattacharyya A,Bhattacharya M,Levine MH,Haldar M
MF48005 was used in Flow cytometry/Cell sorting to uncover a pathway wherein counter-regulation of Spic by NF-κB and STATs attune inflammatory responses and iron metabolism in macrophages.
Tue Jun 30 00:00:00 EDT 2020
The Jeff Mouse Mutant Model for Chronic Otitis Media Manifests Gain-of-Function as Well as Loss-of-Function Effects.
Frontiers in genetics
Kubinyecz O,Vikhe PP,Purnell T,Brown SDM,Tateossian H
MF48005 was used in Immunohistochemistry to investigate the role of FBXO11 and the type of mutation responsible for the phenotype of the Jeff mice.
Wed Mar 17 00:00:00 EDT 2021
3D Co-culture System of Tumor-associated Macrophages and Ovarian Cancer Cells.
Bio-protocol
Long L,Yin M,Min W
MF48005 was used in Flow cytometry/Cell sorting to study how detached ovarian carcinoma cells survive in the peritoneum and form spheroid structures.
Fri Apr 20 00:00:00 EDT 2018
TLR4 signals in B lymphocytes are transduced via the B cell antigen receptor and SYK.
The Journal of experimental medicine
Schweighoffer E,Nys J,Vanes L,Smithers N,Tybulewicz VLJ
MF48005 was used in flow cytometry to show that TLR4 signals through the BCR leading to activation of SYK, ERK, and AKT as well as through MYD88 leading to activation of NFkappaB
Mon May 01 00:00:00 EDT 2017
Microglial-derived microparticles mediate neuroinflammation after traumatic brain injury.
Journal of neuroinflammation
Kumar A,Stoica BA,Loane DJ,Yang M,Abulwerdi G,Khan N,Kumar A,Thom SR,Faden AI
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Wed Mar 15 00:00:00 EDT 2017
Macrophage alternative activation confers protection against lipotoxicity-induced cell death.
Molecular metabolism
Dai L,Bhargava P,Stanya KJ,Alexander RK,Liou YH,Jacobi D,Knudsen NH,Hyde A,Gangl MR,Liu S,Lee CH
MF48005 was used in Flow cytometry/Cell sorting to suggest that ATM cell death is closely linked to metabolic inflammation.
Sun Oct 01 00:00:00 EDT 2017
Respiratory syncytial virus infection of newborn CX3CR1-deficient mice induces a pathogenic pulmonary innate immune response.
JCI insight
Das S,Raundhal M,Chen J,Oriss TB,Huff R,Williams JV,Ray A,Ray P
MF48005 was used in Flow cytometry/Cell sorting to propose that, in infants harbouring mutant CX3CR1, targeting the IL-17R may minimize respiratory syncytial virus severity and hospitalization in early life.
Thu Sep 07 00:00:00 EDT 2017
CXCL16-positive dendritic cells enhance invariant natural killer T cell-dependent IFNγ production and tumor control.
Oncoimmunology
Veinotte L,Gebremeskel S,Johnston B
MF48005 was used in flow cytometry to study the contribution of CXCR6/CXCL16 interactions in glycolipid-dependent invariant natural killer cell activation and tumor control
Wed Jun 01 00:00:00 EDT 2016
Unique role for ATG5 in neutrophil-mediated immunopathology during M. tuberculosis infection.
Nature
Kimmey JM,Huynh JP,Weiss LA,Park S,Kambal A,Debnath J,Virgin HW,Stallings CL
MF48005 was used in flow cytometry to study the role of ATG5 in neutrophil-mediated immunopathology during M. tuberculosis infection
Thu Dec 24 00:00:00 EST 2015
Sphingosine 1-phosphate receptor-1 enhances mitochondrial function and reduces cisplatin-induced tubule injury.
Journal of the American Society of Nephrology : JASN
Bajwa A,Rosin DL,Chroscicki P,Lee S,Dondeti K,Ye H,Kinsey GR,Stevens BK,Jobin K,Kenwood BM,Hoehn KL,Lynch KR,Okusa MD
MF48005 was used in immunohistochemistry - frozen section to test if the protective effect of FTY720 in AKI is mediated by sphingosine 1-phosphate on proximal tubule cells
Wed Apr 01 00:00:00 EDT 2015
The expression of mouse CLEC-2 on leucocyte subsets varies according to their anatomical location and inflammatory state.
European journal of immunology
Lowe KL,Navarro-Núñez L,Bénézech C,Nayar S,Kingston BL,Nieswandt B,Barone F,Watson SP,Buckley CD,Desanti GE
MF48005 was used in flow cytometry to assess surface expression of C-type lectin-like receptor 2 on hematopoietic cells from peripheral blood and secondary lymphoid organs
Tue Sep 01 00:00:00 EDT 2015
Heme-mediated SPI-C induction promotes monocyte differentiation into iron-recycling macrophages.
Cell
Haldar M,Kohyama M,So AY,Kc W,Wu X,Briseño CG,Satpathy AT,Kretzer NM,Arase H,Rajasekaran NS,Wang L,Egawa T,Igarashi K,Baltimore D,Murphy TL,Murphy KM
MF48005 was used in immunohistochemistry (frozen) to investigate how heme regulates Spic in macrophages.
Thu Mar 13 00:00:00 EDT 2014
Lymph node fibroblastic reticular cell transplants show robust therapeutic efficacy in high-mortality murine sepsis.
Science translational medicine
Fletcher AL,Elman JS,Astarita J,Murray R,Saeidi N,D'Rozario J,Knoblich K,Brown FD,Schildberg FA,Nieves JM,Heng TS,Boyd RL,Turley SJ,Parekkadan B
MF48005 was used in Flow cytometry/Cell sorting to characterise a novel population of immune-specialised myofibroblasts derived from lymph node tissue, termed fibroblastic reticular cells.
Wed Aug 13 00:00:00 EDT 2014
MPYS/STING-mediated TNF-α, not type I IFN, is essential for the mucosal adjuvant activity of (3'-5')-cyclic-di-guanosine-monophosphate in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Blaauboer SM,Gabrielle VD,Jin L
MF48005 was used in Flow cytometry/Cell sorting to identify an essential role for STING-mediated TNF-α production in the mucosal adjuvant activity of (3'-5')-cyclic-di-guanosine-monophosphate in vivo and reveal a novel interferon-I stimulation-independent STING-NF-κB-TNF-α pathway.
Wed Jan 01 00:00:00 EST 2014
Cranial irradiation alters the brain's microenvironment and permits CCR2+ macrophage infiltration.
PloS one
Morganti JM,Jopson TD,Liu S,Gupta N,Rosi S
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Mon Feb 02 00:00:00 EST 2015
Sonic Hedgehog contributes to gastric mucosal restitution after injury.
Laboratory investigation; a journal of technical methods and pathology
Xiao C,Feng R,Engevik AC,Martin JR,Tritschler JA,Schumacher M,Koncar R,Roland J,Nam KT,Goldenring JR,Zavros Y
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Sonic Hedgehog contributes to gastric mucosal restitution after injury.
Laboratory investigation; a journal of technical methods and pathology
Xiao C,Feng R,Engevik AC,Martin JR,Tritschler JA,Schumacher M,Koncar R,Roland J,Nam KT,Goldenring JR,Zavros Y
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Human and mouse macrophages collaborate with neutrophils to kill larval Strongyloides stercoralis.
Infection and immunity
Bonne-Année S,Kerepesi LA,Hess JA,O'Connell AE,Lok JB,Nolan TJ,Abraham D
MF48005 was used in Flow cytometry/Cell sorting to demonstrate a novel function by which alternatively activated macrophages control large extracellular parasites.
Sun Sep 01 00:00:00 EDT 2013
Chlamydia psittaci genetic variants differ in virulence by modulation of host immunity.
The Journal of infectious diseases
Miyairi I,Laxton JD,Wang X,Obert CA,Arva Tatireddigari VR,van Rooijen N,Hatch TP,Byrne GI
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2011
Chlamydia psittaci genetic variants differ in virulence by modulation of host immunity.
The Journal of infectious diseases
Miyairi I,Laxton JD,Wang X,Obert CA,Arva Tatireddigari VR,van Rooijen N,Hatch TP,Byrne GI
Published figure using F4/80 monoclonal antibody (Product # MF48005) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2011
Toll-like receptor 2 (TLR2) plays a major role in innate resistance in the lung against murine Mycoplasma.
PloS one
Love W,Dobbs N,Tabor L,Simecka JW
MF48005 was used in flow cytometry to study the role of TLR2 signaling during Mycoplasma pulmonis infection
Thu May 20 00:00:00 EDT 2010
Toxoplasma gondii prevents chromatin remodeling initiated by TLR-triggered macrophage activation.
Journal of immunology (Baltimore, Md. : 1950)
Leng J,Butcher BA,Egan CE,Abi Abdallah DS,Denkers EY
MF48005 was used in flow cytometry to study the effect of Toxoplasma gondii on chromatin remodeling
Thu Jan 01 00:00:00 EST 2009
Eosinophils develop in distinct stages and are recruited to peripheral sites by alternatively activated macrophages.
Journal of leukocyte biology
Voehringer D,van Rooijen N,Locksley RM
MF48005 was used in flow cytometry to report distinct stages of eosinophil development using transgenic mice
Fri Jun 01 00:00:00 EDT 2007
The p47 GTPases Iigp2 and Irgb10 regulate innate immunity and inflammation to murine Chlamydia psittaci infection.
Journal of immunology (Baltimore, Md. : 1950)
Miyairi I,Tatireddigari VR,Mahdi OS,Rose LA,Belland RJ,Lu L,Williams RW,Byrne GI
MF48005 was used in flow cytometry to report that Irgb10 and Iigp2 confer differential susceptibility against murine chlamydial infection
Wed Aug 01 00:00:00 EDT 2007
Helminth-modified pulmonary immune response protects mice from allergen-induced airway hyperresponsiveness.
Journal of immunology (Baltimore, Md. : 1950)
Mangan NE,van Rooijen N,McKenzie AN,Fallon PG
MF48005 was used in flow cytometry to elucidate the mechanisms that regulate the inverse relationship between helminth infections and allergies
Sun Jan 01 00:00:00 EST 2006
Effect of thermal injury on splenic myelopoiesis.
Shock (Augusta, Ga.)
Noel JG,Guo X,Wells-Byrum D,Schwemberger S,Caldwell CC,Ogle CK
MF48005 was used in flow cytometry to measure the number of splenic myeloid progenitors and their progeny after burn injury
Tue Feb 01 00:00:00 EST 2005