CD16/CD32 Monoclonal Antibody (FRC-4G8) - FAQs
CD16/CD32 Monoclonal Antibody (FRC-4G8) - FAQs
View additional product information for CD16/CD32 Monoclonal Antibody (FRC-4G8) - FAQs (MFCR004)
1 product FAQs found
What should I use to block my cells for flow cytometry analysis?
Use serum from the same species as the host species of the secondary antibody for blocking. If the serum is not available, use from 2 to 5% BSA (Fraction V, defatted). If using only a primary antibody, such as directly-labeled mouse primary antibodies, a good blocking reagent is Fc block. CD16 + CD32 Antibody (FRC-4G8) (Cat. No. MFCR004 or MA5-16680) is a low-affinity receptor for the Fc region of immunoglobulin gamma complexes.