CD30 / TNFRSF8 Antibody, RPE conjugate (HRS-4)

Catalog number: MHCD3004

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD30 / TNFRSF8 Monoclonal Antibody for Flow
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Reactivity: Human
Validated Application: Flow Cytometry
Host Species: Mouse
Clonality: Monoclonal
Clone ID: HRS-4
Target Gene: CD30
Gene ID: 943
Label or Dye: R-PE
Excitation⁄Emission (nm): 488⁄575
Number of Reactions: 100 Tests
Product Size: 0.5 mL
Research Category: Signal Transduction
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products