iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
Thermo Scientific™
iConnect Split Splitless (SSL) Injector Module with Integrated Backflush for TRACE 1600 Series GC
Manage a difficult and heavy matrix through integrated backflushing, protect from contamination, increase throughput and decrease running costs.
Have Questions?
Número do catálogoQuantity
MI-190000-0019Each
Número do catálogo MI-190000-0019
Preço (BRL)
-
Quantity:
Each
Extend the life of your analytical column and maximize laboratory throughput by eliminating high boiling compounds. The Thermo Scientific iConnect SSL-BKF injector module enables you to add pre-column, post-column, or mid-column backflush to your split or splitless injection.
Thanks to the self-adjusting pressure during backflushing, the integrated pneumatic control eliminates complexity and the cost of an additional auxiliary gas module, and facilitates method setup.
Save analysis time and gain in robustness against heavy contamination. Experience the simplicity of backflush injection to:
  • Extend instrument uptime and reduce maintenance by preventing column and detector contamination through removal of unwanted high boiling compounds
  • Confidently select the best backflush configuration for your sample composition, with correct and self-adjusted pneumatic settings
  • Quickly and easily install the iConnect SSL-BKF module, thanks to the modular design of the TRACE 1600 Series GC
  • Set up your columns connection with the use of the Thermo Scientific™ microfluidic device, based on SGE's SilFlow® technology, easily accessible, fitting neatly within the column oven
  • Control all the injection steps through the Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software
Especificações
DescriptioniConnect Split/Splitless (SSL) injector module with integrated Backflush
QuantityEach
For Use With (Equipment)Trace 1600 Series
Injection ModeSplit and Splitless injection with reversal flow (backflush)
TypeSSL Injector with integrated backflush
Unit SizeEach