HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
Thermo Scientific™
HeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
Easily convert your iConnect SSL to save significantly on carrier gas consumption, whether using helium or hydrogen, and maintain your current GC methods.
Have Questions?
Catalog NumberInjection Mode
MI-190000-0022Split/Splitless
Catalog number MI-190000-0022
Price (TWD)
-
Injection Mode:
Split/Splitless
Improve the sustainability of your laboratory, regardless of whether helium or hydrogen is your carrier gas. To significantly reduce carrier gas consumption while running samples and when the GC is idle, the Thermo Scientific HeSaver-H2Safer kit easily transforms the iConnect SSL injector module. To save time and cost for re-validation, existing methods remain unaffected. When helium is used as carrier, the HeSaver mode substantially extends cylinder lifetime and mitigates helium shortage concerns. When hydrogen is used as carrier, the H2Safer mode ensures safer operations and optimized use of hydrogen generators.
Maintain your existing methods and dramatically save on carrier gas consumption, whether using helium or hydrogen carrier gas:
For laboratories using helium as carrier gas
  • Mitigate helium shortage concerns, including delayed delivery and increased costs
  • Maintain existing validated methods
  • Achieve optimal GCMS performance with no compromise on sensitivity or spectra matching
For laboratories using hydrogen as carrier gas
  • Benefit from safer operating conditions thanks to a limited max flow rate, removing the need for hydrogen sensor in the GC oven
  • Serve more GCs with a hydrogen generator
  • Ensure inert injection environment in the hot SSL since hydrogen is not flushing into the injector
Additional benefits of using the HeSaver-H2Safer option
  • Save time for inlet maintenance. The modified SSL injector can be accessed while the carrier gas flows into the column, removing the need to cool the GC oven.
  • Reduce contamination from the injector into the column. The carrier gas does not flow through the injector, which protects the column from common contaminants (septum/sample matrix/by-products).
Specifications
DescriptionHeSaver-H2Safer™ kit for iConnect™ SSL injector module TRACE™ 1600 Series GC
QuantityEach
For Use With (Equipment)iConnect SSL injector on TRACE 1600 Series GC. Compatible with capillary columns from 50 µm to 320 µm inner diameter
Injection ModeSplit/Splitless
Product TypeKit for iConnect SSL injector module
Unit SizeEach