What type of slide is used for the NCode microRNA microarrays?
NCode miRNA Microarrays are generally contact-printed using microquill pins on Corning Epoxide-Coated glass slides.
Search
View additional product information for NCode™ Human miRNA Microarray V3 - FAQs (MIRAH305)
1 product FAQs found