MCA-2 M-Prep Conditioner A, Measurement Group™
MCA-2 M-Prep Conditioner A, Measurement Group™
MCA-2 M-Prep Conditioner A, Measurement Group™
Measurement Group

MCA-2 M-Prep Conditioner A, Measurement Group™

A mild phosphoric-acid for surface preparation.
Have Questions?
Número de catálogo MMF006557
Precio (USD)
-
Pedido a granel o personalizado
Especificaciones
Punto de ebullición158°C
CAS7664-38-2
Color≤10 APHA
Punto de fusión40°C
Fórmula molecularH3O4P
Ver más
A mild phosphoric-acid for surface preparation.

A mild phosphoric-acid for surface preparation.

  • Mild etchant to accelerate the surface cleaning process
  • Immediately followed by M-Prep Neutralizer 5A