MAGPIX™ Performance Verification Kit
Invitrogen™

MAGPIX™ Performance Verification Kit

The convenient performance verification kit contains xMAP™ verification reagents that are used to verify calibration of the MAGPIX™ classification andRead more
Catalog NumberQuantity
MPXPVERK2525 reactions
Catalog number MPXPVERK25
Price (GBP)
574.00
Each
Quantity:
25 reactions
Price (GBP)
574.00
Each
The convenient performance verification kit contains xMAP™ verification reagents that are used to verify calibration of the MAGPIX™ classification and reporter channels and fluidic integrity. Verification uses system controls to help ensure that the analyzer is functioning properly with current calibration settings. The kit is intended to be used with the off-plate reagent block. Each kit provides 25 verifications.

Each kit includes:

Stripwells

• A lot-specific CD for importing target values

The MAGPIX™ system is a versatile, compact, fluorescent detection system based on Luminex's proven xMAP™ technology. The system is a simple immunoassay platform designed for the simultaneous analysis of up to 50 intracellular and extracellular markers in single biological samples. The MAGPIX™ system provides users with a powerful multiplexing tool that offers higher sample throughput, increased assay flexibility, reduced sample volume, and lower costs than traditional ELISA and Western blotting.

For research use only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex Instruments, MAGPIX System
Product LineMAGPIX
Quantity25 reactions
Recommended StorageThe MAGPIX™ Performance Verification Kit includes:

• 25 strip wells (PN: 13-52047)
• MAGPIX™ Verifier Microspheres, 5 mL (PN MPXVER-05)
• MAGPIX™ Fluidics1 Microspheres, 5 mL (PN MPXFLUID1-05)
• MAGPIX™ Fluidics2 Microspheres, 5 mL (PN MPXFLUID2-05)
• MAGPIX™ Performance Verification Kit CD (PN 89-20234-00-001)

Store at 2-8°C
Shipping ConditionWet Ice
Unit SizeEach

Frequently asked questions (FAQs)

How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Impact of rewarming rate on interleukin-6 levels in patients with shockable cardiac arrest receiving targeted temperature management at 33 °C: the ISOCRATE pilot randomized controlled trial.
Authors:Lascarrou JB, Guichard E, Reignier J, Le Gouge A, Pouplet C, Martin S, Lacherade JC, Colin G
Journal:Crit Care
PubMed ID:34920723
While targeted temperature management (TTM) has been recommended in patients with shockable cardiac arrest (CA) and suggested in patients with non-shockable rhythms, few data exist regarding the impact of the rewarming rate on systemic inflammation. We compared serum levels of the proinflammatory cytokine interleukin-6 (IL6) measured with two rewarming rates ... More