Thermo Scientific™
General-Purpose (GP) Series Lab Refrigerators
General-Purpose (GP) Series Lab Refrigerators
Related applications:
Cold Storage
Maintain daily lab applications with the help of Thermo Scientific™ General-Purpose (GP) Series Refrigerators, which feature digital controls and excellent temperature stability.
Back to top
Performance:
  • Digital temperature controller with intuitive interface
  • Excellent temperature stability of ±3°C
  • High-density, fluorine free insulation
  • Automatic defrost
Security:
  • Optional chart recorder
  • High/Low temperature, visual/audio alarms
  • Choice of solid or glass lockable doors (refrigerators only)
  • Easily accessible contacts for backup monitoring
  • Door ajar alarm (except sliding glass doors)
  • Control panel with key lock
Ordering Information:
  • For 115V 60Hz.
  • Equipped with NEMA 5-15P line cord plug; require NEMA 5-15R receptacle.
  • Warranty varies by country, please contact us for details