Biotin Ab-2, Mouse Monoclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Detect both the free and protein-conjugated forms of biotin with Thermo Scientific™ Biotin Ab-2, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

Back to top

Description

Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Host Species: Mouse

Clone: BTN.4

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Biotin conjugated to BSA

Recommended for:

  • Western Blotting
  • Confocal Microscopy
  • Flow Cytometry
  • Image Analysis
  • In situ Nucleic Acid Hybridization

Note:

USA: RUO; Int'l: RUO



Documents

Manuals & protocols