Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: LAM03 + HP6054

Isotype: IgG1/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO



Documents

Manuals & protocols