Thermo Scientific™

JAK3 Ab-1, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Thermo Scientific™ JAK3 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

JAK3 (also designated L-JAK) is a member of the Janus family of protein tyrosine kinases along with JAK1, JAK2, or Tyk2.. Subsequent to activation, these kinases function to activate various members of the Stat family of transcription factors by phosphorylation on critical tyrosine regulatory sites. JAK3 is activated in response to IL-3 and IL-4 in T cells and myeloid cells. JAK3 activation requires the serine-rich membrane-proximal region of the IL-2 receptor β-chain, but does not require the acidic domain that is required for association and activation of Src family kinases. Expression of JAK3 is most commonly found in NK and T cells.

Host Species: Mouse

Clone: A1-14-16

Isotype: IgG2a/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein encoding C-terminus corresponding to aa 1076-1100 of murine JAK3

Molecular Weight: 115kDa

Positive Control: Jurkat cells; Tonsil, Pancreas

Cellular Localization: Membrane, cytoplasmic and occasionally nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

General References:

1. Blood 87: 3151-3160 (1996).



Documents

Manuals & protocols