Thermo Scientific™

Lab Vision™ LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Luteinizing hormone (LH) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common α-subunit but also a spesofic β-subunit, which characterizes each of these hormones.

Host Species: Mouse

Clone: LH01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human LH-β

Molecular Weight: 14kDa (β) and 28kDa (αβ)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Ab-1 recognizes the beta subunit both free and intact of luteinizing hormone (LH). Ab-1 is useful for the labeling of normal gonadotropic cells of the pituitary and also for the classification of pituitary adenomas and in the differential identification of primary and metastatic tumors of the pituitary.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Documents

Manuals & protocols