Thermo Scientific™

Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody

 Related applications: Anatomical Pathology

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Host Species: Mouse

Clone: MAC 387

Isotype: IgG1

Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.

Epitope: Not determined

Immunogen: Affinity purified monocyte membrane preparation

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Coleman N Stanley MA. et al. Human Pathology, 1994, 25(1):73-9.

2. Andersen CB et al. Apmis, 1994, 102(1):23-37.



Documents

Manuals & protocols