Thermo Scientific™

Lab Vision™ CD4 Ab-8, Mouse Monoclonal Antibody

Related applications:

Anatomical Pathology

Obtain accurate and reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD4 Ab-8, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Back to top
CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV.

Host Species: Mouse

Clone: 4B12

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 59 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Stein H, et. al. (1984) Adv Cancer Res, 42:67-147.