Thermo Scientific™

Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology

Test for macrophages and monocytes with Thermo Scientific™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

CD68 glycoprotein is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: PG-M1

Isotype: IgG3/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Gaucher cells

Molecular Weight: 110 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Fallini B, et al. (1993) Am J Pathol, 142:1359.

2. Horny H P, et al. (1992) Hum Pathol, 24:355.



Documents

Manuals & protocols