Thermo Scientific™

Lab Vision™ Hepatocyte Ab-1, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology

Obtain accurate, reproducible results in Immunohistochemistry experiments with Thermo Scientific™ Hepatocyte Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Host Species: Mouse

Clone: OCH1E5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Formalin-fixed failed human allograft liver that was mechanically disrupted

Positive Control: Liver CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Minervini M I, et al. (1997) Mod Pathol, 10:686-692.

2. Murakata L A, et al. (2000) Mod Pathol, 13:874-881.

3. Fasano M, et al. (1998) Mod Pathol, 11:934-938.

4. Wennerberg A E, et al. (1993) Am J Pathol, 11:1050-1054.



Documents

Manuals & protocols