Thermo Scientific™

Lab Vision™ CA19-9/Sialyl Lewis a (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Host Species: Mouse

Clone: 116-NS-19-9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Sialic acid-containing residue of the monosialoganglioside

Immunogen: Human colon carcinoma cell line SW1116

Positive Control: Colon CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

General References:

1. Kaprowski H, et al. (1979) Somatic Cell Genet, 5:957.

2. Magnani J L, et al. (1882) J Biol Chem, 257:14635.

3. Atkinson B F, et al. (1982) Canc res, 42:4820.

4. Hanai N, et al. (1990) Anti Canc Res, 10:1579.



Documents

Manuals & protocols