CA 15-3™ Ab-1, Mouse Monoclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CA 15-3 Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

Back to top

Description

Host Species: Mouse

Clone: DF3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Glycosidic linkage between carbohydrate and protein moieties

Immunogen: Membrane enriched fractions of human metastatic breast carcinoma

Molecular Weight: 300-450kDa

Positive Control: Ovarian CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Kufe D, et al. (1984) Hybridoma, 3:223.

2. Abe M, et al. (1987) J Immunol, 139:257.

3. Sekine H, et al. (1985) J Clin Oncol, 3:1355.

4. Friedman et al. (1986) Cancer Res, 46:5189.



Documents

Manuals & protocols