Thermo Scientific™

Lab Vision™ Glycophorin A Ab-4, Mouse Monoclonal Antibody

Related applications:

Anatomical Pathology

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Glycophorin A Ab-4, Mouse Monoclonal Antibody.
Back to top
Glycophorin A is a sialoglycoprotein present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus.

Host Species: Mouse

Clone: JC159

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Molecular Weight: 39 kDa

Positive Control: Human erythrocytes in placenta

Cellular Localization: Membrane Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Greaves M F, et al. (1983) Blood, 61:645.

2. Loken M R, et al. 91987) Blood, 69:255.

3. Erber W N, et al. (1991) Hematol Rev, 5:113.