Cyclin D3 Ab-1, Mouse Monoclonal Antibody

Thermo Scientific™  Related applications: Anatomical Pathology

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications with Thermo Scientific™ Cyclin D3 Ab-1, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}

Introducing shared lists
Back to top


Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.

Host Species: Mouse

Clone: DCS-22

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: LS174T cells, Breast CA or tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry


USA: RUO; Int'l: RUO

General References:

1. Bartkova J, et. al. Cancer Res, 1995, 55:949-5

2. Lukas J et al. Mol Cell Biology, 1995, 15(5):2600-11.


Manuals & protocols