Thermo Scientific™

Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-11, Mouse Monoclonal Antibody

Related applications:

Anatomical Pathology

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-11, Mouse Monoclonal Antibody.
Back to top
EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 199.12

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Extracellular domain of human recombinant EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native only)
  • Effect on EGF Binding (Noninhibitory)

Note:

USA: RUO; Int'l: RUO

General References:

1. Sato JD, et. al. Mol Biol Med, 1983, 1:511-29.

2. den Hartigh JC, et. al. Biochimica Biophys Acta, 1993, 1148:249-56.

3. Kawamoto T, et. al. Proceedings of the National Academy of Sciences, USA, 1983, 80:1337-41.