Thermo Scientific™

Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

Host Species: Mouse

Clone: COL-1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Immunogen: Human colon carcinoma extract

Molecular Weight: 180 kDa

Positive Control: LS174T cells, Colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

General References:

1. Shi ZR, et. al. J Histo Cyto, 1994, 42(9):1215-9.

2. Myers RB, et. al. J Urology, 1994, 152(1):243-6.

3. Ohuchi N, et. al. Cancer Res, 1987, 47(13):3565-71.

4. Shimada S, et. al. Cancer Res, 1991, 51:5694-703.

5. Robbins PF, et. al. Int J Cancer, 1993, 53(6):892-7.

6. Siler K, et. al. Biotech Ther 1993, 4(3-4):163-81.

7. Muraro R, et. al. Cancer Research, 1985, 45:5769-80.



Documents

Manuals & protocols