Thermo Scientific™

Lab Vision™ Insulin Receptor (β -Subunit) Ab-6, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Recommended for a variety molecular biology techniques, Thermo Scientific™ Insulin Receptor (β-Subunit) Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: CT-3

Isotype: IgG1

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: Recombinant fragment including the C-terminal 100 amino acids of human insulin receptor

Molecular Weight: 95kDa

Positive Control: HEP-G-2, IM-9 lymphocyte cells; Placenta or brain

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Affinity Purification (With peptide elution)
  • ELISA (As capture antibody only)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO



Documents

Manuals & protocols