Thermo Scientific™

Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody

Related applications:

Anatomical Pathology

Ideal for Western Blotting, Thermo Scientific™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Back to top
Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.

Host Species: Mouse

Clone: SBF5

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human HGF protein

Molecular Weight: ∽82kDa

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO