SILAC Protein ID & Quantitation Media Kit, with Lysine and IMDM-Flex - FAQs
SILAC Protein ID & Quantitation Media Kit, with Lysine and IMDM-Flex - FAQs
View additional product information for SILAC Protein ID & Quantitation Media Kit, with Lysine and IMDM-Flex - FAQs (MS10032)
1 product FAQs found
Are the heavy amino acids included in the price of the SILAC kits?
Yes, the kit includes all the components for labeling cells with the heavy lysine. Heavy arginine is sold as a separate item (Cat. No. MS10009) for researchers who want to perform a double-label experiment using heavy lysine and arginine.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.