Product Image
Gibco™

Mouse (CD-1) Microsomes

Liver microsomes prepared from a pool of male CD-1 mice. Total protein concentration: 20mg/mL.
Have Questions?
카탈로그 번호수량
MSMCPL0.5 mL
카탈로그 번호 MSMCPL
제품 가격(KRW)
120,000
온라인 행사
Ends: 30-Sep-2026
133,000
할인액 13,000 (10%)
Each
수량:
0.5 mL
제품 가격(KRW)
120,000
온라인 행사
Ends: 30-Sep-2026
133,000
할인액 13,000 (10%)
Each
Liver microsomes prepared from a pool of male CD-1 mice.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.
사양
농도20 mg/mL
형식Tube
제품라인Gibco
제품 유형Mouse Microsome
순도Research Grade
수량0.5 mL
배송 조건Dry Ice
세포하위구조 국지화Microsomes
공여자 소스Pooled
성별Male
숙주 종Mouse
물리적 형태Frozen
TissueLiver
Unit SizeEach
구성 및 보관
Store between -60°C and -85°C.