VetMAX™ M. tuberculosis Complex PCR Kit
Applied Biosystems™

VetMAX™ M. tuberculosis Complex PCR Kit

The VetMAX M. tuberculosis Complex PCR Kit is a single-well, duplex, real-time PCR assay for the detection of all mycobacteriaRead more
Have Questions?
Catalog NumberQuantity
MTBC100 reactions
Catalog number MTBC
Price (USD)
-
Quantity:
100 reactions
The VetMAX M. tuberculosis Complex PCR Kit is a single-well, duplex, real-time PCR assay for the detection of all mycobacteria belonging to the Mycobacterium tuberculosis Complex in bovine lymph nodes and surrounding tissues. The kit is a highly sensitive and specific test for the detection of bovine tuberculosis, allowing reliable and rapid screening, exclusion of non-infected animals, and confirmation of infected animals.

All components needed for the PCR reaction are included in the kit:
• Mix MTBC: contains TaqMan primers and probes, buffer, and enzyme for optimized duplex real-time PCR amplification of targets
• EPC MTBC: DNA template containing the M. tuberculosis Complex target sequence (external positive control). It serves as a positive control for the real-time PCR components and is used to set the threshold cycle (Ct) for evaluating test results
• IPC MTBC: added to each test and control sample at the lysis step of the DNA isolation procedure (exogenous internal positive control). The IPC serves as a control for the DNA isolation procedure and is used to monitor for the presence of PCR inhibitors

The VetMAX M. tuberculosis Complex PCR Kit is part of a workflow solution with the MagVet Universal Isolation Kit (Cat. No. MV384) and the Kingfisher Flex Magnetic Particle Processor or Kingfisher mL Flex Magnetic Particle Processor (Cat. Nos. 5400630 and 5400640).

Regulatory requirements vary by country; products may not be available in your geographic area.
Specifications
FormatTube
No. of Reactions100 reactions
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 reactions
Sample TypeBacteria
Shipping ConditionDry Ice
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR Method2-step RT-qPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Store at -5° to -30°C.

Frequently asked questions (FAQs)

What are the probes included in the VetMAX M. tuberculosis Complex PCR Kit?

The TaqMan assay for TUB is labeled with FAM and quenched by NFQ (non-fluorescent quencher). The TaqMan assay for IPC is labeled with VIC and quenched by NFQ (non-fluorescent quencher).