Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »
Zeiss 40X Objective, LD A-Plan (0.5 NA)
Catalog number: N010097
Thermo Scientific™  Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Zeiss™ 40X (0.5NA) LD A-Plan objective lens for the Thermo Scientific™ ArrayScan™ HCA Readers is made by the world renowned optics company, Zeiss. The Zeiss 40X (0.5NA) LD A-Plan lens represents the finest in optical with a magnification power suited for subcellular imaging, and a numerical aperture which strikes a balance between resolution and versatility.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): ArrayScan HCA Readers
Lens Type: Dry
Magnification: 40x (0.5NA)
Product Line: Zeiss
Product Size: 1 each
Type: Objective Lens
Documents
Manuals & protocols