Thermo Scientific™
Zeiss 20X Objective, Plan-Apochromat (0.8 NA)
Catalog number:  N010098
 Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Zeiss™ 20X (0.8NA) Plan-Apochromat objective lens for the Thermo Scientific™ ArrayScan™ HCA Readers is made by the world renowned optics company, Zeiss. The Zeiss 20X (0.8NA) Plan-Apochromat lens represents the finest in optical with a magnification power suited for subcellular imaging, and a numerical aperture which strikes a balance between resolution and versatility.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): ArrayScan HCA Readers
Lens Type: Dry
Magnification: 20x (0.8NA)
Product Line: Zeiss
Product Size: 1 each
Type: Objective Lens
Documents
Manuals & protocols