ArrayScan™ Brightfield Module
ArrayScan™ Brightfield Module
Thermo Scientific™
ArrayScan™ Brightfield Module
The Thermo Scientific™ ArrayScan™ Brightfield Module is an affordable option that adds white light illumination to high content screening (HCS)자세히 알아보기
Have Questions?
카탈로그 번호수량
N010115A1 unit
카탈로그 번호 N010115A
제품 가격(KRW)
-
수량:
1 unit
The Thermo Scientific™ ArrayScan™ Brightfield Module is an affordable option that adds white light illumination to high content screening (HCS) experiments. It allows imaging of unlabeled samples to quantify morphological features and the resulting data can be interleaved with fluorescence data from the same sample. The ArrayScan Brightfield Module has integrated software control to capture, focus, and analyze transmitted light images at single or multiple timepoints.
Features of the ArrayScan Brightfield Module :
• Capture, analyze, and visualize transmitted light images interleaved with fluorescence images
• Label-free detection
• Autofocus in transmitted light mode permitting imaging on multiple plate types, cell lines and magnifications
• Color composites can be used to verify location of fluorescent targets within a cell or organism
The ArrayScan Brightfield Module is fully integrated with the ArrayScan HCA Reader software (Thermo Scientific™ HCS Studio™ Cell Analysis Software) enabling the capture, analysis, and visualization of transmitted light images interleaved with fluorescence images at single or multiple time points.
For Research Use Only. Not for use in diagnostic procedures.
사양
색상None
검출 방법Brightfield
치수170 mm x 50mm x 170mm
용도(장비)ArrayScan HCA Readers
작동 환경Permissible ambient temperature range during operation : 18°C to 25°C, Permissible relative humidity during operation (non-condensing): >75% at 30°C
옵틱스LED Illumination
수량1 unit
소프트웨어 호환성Thermo Scientific™ HCS Studio Cell Analysis Software
전압100-240 VAC
중량(미터법)1 kg
제품라인ArrayScan
Unit Size1 unit