Thermo Scientific™

ArrayScan™ Liquid Handling Module

Catalog number: N010117B
Thermo Scientific™

ArrayScan™ Liquid Handling Module

Catalog number: N010117B
Catalog Number
N010117B
Unit Size
1 each
Price (USD)
Catalog NumberUnit SizePrice (USD)
N010117B1 eachRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ ArrayScan™ Liquid Handling Module is designed to work with the Thermo Scientific™ ArrayScan™ Live Cell Module, expanding the flexibility of the platform for analysis of complex cellular states of cells. The ArrayScan Liquid Handling Module has functionalities to aspirate, dispense, and mix, from four deck locations. The module is capable of single-tip aspirate and dispense at any location, creating a highly flexible instrument. Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the ArrayScan Liquid Handling Module utilizes disposable tips with a dispense volume range of 3 to 200μL.

Features of the ArrayScan Liquid Handling Module:

• Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the module utilizes disposable tips with a dispense volume range of 3 to 200μL
• Full function liquid handling from any plate or trough (3 plate and 1 quarter trough)
• Industry standard arm with single pipette tip driven by syringe pump (e.g., aspirate, dispense)
• Compatible with all ArrayScan Modules
• Dynamic access to compound and assay plate
• One-to-many and many-to-one transfer scenarios are supported

Includes:

ArrayScan Liquid Handling Module : Automated, single-tip, liquid handling robot module for the Thermo Scientific™ ArrayScan™ High Content Analysis (HCA) Readers using disposable tips (standard and liquid level sensing) to both aspirate and dispense from multiple deck and live cell chamber locations.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
ArrayScan
For Use With (Equipment)
ArrayScan HCA Readers
Dispense Volume
3 to 200 μL (± 5% in 1 μL increments)
Format
6-wells, 8-wells, 384-well plate, 24-well plate, 96-well plate
Software Compatibility
Thermo Scientific™ HCS Studio Cell Analysis Software
Operating Environment
Permissible ambient temperature range during operation : + 18 deg C to + 25 deg C, Permissible relative humidity during operation (non-condensing): > 75% (at 30 deg C)
Dimensions
87.63 cm (34.5 in.) (W) x 109.22 cm (43 in.) (H) x 74 cm (29 in.) (D)
Weight (Metric)
59 kg
Voltage
100 V to 240 V
Wattage
150 W (+ 500 W from ArrayScan Reader)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text