REAGENT RESERVOIR 250ML NON ST - FAQs
REAGENT RESERVOIR 250ML NON ST - FAQs
View additional product information for REAGENT RESERVOIR 250ML NON ST - FAQs (N07497)
1 product FAQs found
Can the Multidrop Combi nL reagent reservoir be autoclaved?
Yes, the reagent reservoir can be autoclaved.