The aromatic dialdehyde naphthalene-2,3-dicarboxaldehyde (NDA) is essentially nonfluorescent until reacted with a primary amine in the presence of excess cyanideRead more
Have Questions?
Catalog Number
Quantity
N1138
100 mg
Catalog number N1138
Price (USD)
187.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
187.00
Each
The aromatic dialdehyde naphthalene-2,3-dicarboxaldehyde (NDA) is essentially nonfluorescent until reacted with a primary amine in the presence of excess cyanide or a thiol, to yield a fluorescent isoindole with excitation/emission maxima ∼419/493 nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Label or DyeNDA (naphthalene-2,3-dicarboxaldehyde)
Quantity100 mg
Shipping ConditionRoom Temperature
TargetPrimary Amine
FormSolid
TypeNaphthalene-2,3-Dicarboxaldehyde (NDA)
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.
Citations & References (140)
Citations & References
Abstract
Electrochemical detection in microcolumn separations.
Authors:Ewing AG, Mesaros JM, Gavin PF
Journal:Anal Chem
PubMed ID:8017638
Derivatization in capillary electrophoresis.
Authors:Bardelmeijer HA, Lingeman H, de Ruiter C, Underberg WJ
Journal:J Chromatogr A
PubMed ID:9646486
'In recent years capillary electrophoresis (CE) has been developed into a versatile separation technique, next to gas and liquid chromatography (LC), well suited for the determination of a wide variety of e.g., pharmaceutical, biomedical and environmental samples. The main advantages of CE over chromatographic separation techniques are its simplicity and ... More
Focusing of nitric oxide mediated nitrosation and oxidative nitrosylation as a consequence of reaction with superoxide.
Authors:Espey MG, Thomas DD, Miranda KM, Wink DA
Journal:Proc Natl Acad Sci U S A
PubMed ID:12177414
'The impact of nitric oxide (NO) synthesis on different biological cascades can rapidly change dependent on the rate of NO formation and composition of the surrounding milieu. With this perspective, we used diaminonaphthalene (DAN) and diaminofluorescein (DAF) to examine the nitrosative chemistry derived from NO and superoxide (O2-) simultaneously generated ... More
Tumor cell-derived nitric oxide is involved in the immune-rejection of an immunogenic murine lymphoma.
Authors:Hu DE, Dyke SO, Moore AM, Thomsen LL, Brindle KM
Journal:Cancer Res
PubMed ID:14729619
'The roles played by host-derived nitric oxide (NO) in the growth and subsequent immune rejection of a immunogenic murine lymphoma were investigated by growing the tumor in mice in which the gene for either inducible NO synthase (iNOS) or endothelial NOS (eNOS) had been ablated. This showed that NO from ... More
Biological activity of nitric oxide in the plasmatic compartment.
Authors:Wang X, Tanus-Santos JE, Reiter CD, Dejam A, Shiva S, Smith RD, Hogg N, Gladwin MT
Journal:Proc Natl Acad Sci U S A
PubMed ID:15258287
'There exist reaction products of nitric oxide (NO) with blood that conserve its bioactivity and transduce an endocrine vasomotor function under certain conditions. Although S-nitrosated albumin has been considered the major species subserving this activity, recent data suggest that additional NO species, such as nitrite, nitrated lipids, N-nitrosamine, and iron-nitrosyl ... More