Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip
Actual product may vary
Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip
Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip
Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip
Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip
Life Technologies Holdings Pty Ltd

Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip

Enables precise small-volume dispensing, ensuring accurate reagent handling for sensitive laboratory applications.
Have Questions?
Catalog NumberVolume (Metric)
N227000.5 to 50 μL
Catalog number N22700
Price (USD)
1,340.10
Online exclusive
1,504.00
Save 163.90
Each
Volume (Metric):
0.5 to 50 μL
Price (USD)
1,340.10
Online exclusive
1,504.00
Save 163.90
Each
Designed for use with compatible dispensing systems to deliver reliable, low-volume liquid handling. It supports reproducibility and accuracy in workflows requiring precise reagent distribution.
  • Optimized for precise low-volume reagent dispensing
  • Compatible with SMART+ dispensing systems
  • Ensures accurate delivery for sensitive assays and workflows
  • Supports consistent and reproducible results
  • Easy installation, removal, and maintenance
  • Ideal for applications requiring minimal reagent usage
Specifications
AutoclavableAutoclavable
DescriptionSMART+ Small Plastic Tip
For Use With (Application)Automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA.
For Use With (Equipment)Multidrop Combi SMART+
Quantity1 Ea.
Volume (Metric) Dead<1 mL
MaterialIn contact with liquid: PEEK, silicone, and PP (polypropylene)
Product TypeTube Dispensing Cassette
Volume (Metric)0.5 to 50 μL
Unit SizeEach

Frequently asked questions (FAQs)

What are the tubing lengths of the regular length and extended length cassettes for the Multidrop Combi Reagent Dispensers?

Regular length tubing (small and standard) cassette length is 40 cm long.

Extended length tubing (small and standard) cassette length is 200 cm long.
Note: You can cut the extended length tubing to the desired length but we recommend checking the calibration on the cassette, once modified, to ensure it is providing accurate results.