Multidrop Combi SMART+ Small Tube Cassette, Metal Tip
Actual product may vary
Multidrop Combi SMART+ Small Tube Cassette, Metal Tip
Multidrop Combi SMART+ Small Tube Cassette, Metal Tip
Multidrop Combi SMART+ Small Tube Cassette, Metal Tip
Multidrop Combi SMART+ Small Tube Cassette, Metal Tip
Life Technologies Holdings Pty Ltd

Multidrop Combi SMART+ Small Tube Cassette, Metal Tip

Delivers consistent small-volume dispensing, helping laboratories achieve reliable and reproducible results.
Have Questions?
Catalog NumberVolume (Metric)
N227020.5 to 50 μL
Catalog number N22702
Price (USD)
1,340.10
Online exclusive
1,504.00
Save 163.90
Each
Volume (Metric):
0.5 to 50 μL
Price (USD)
1,340.10
Online exclusive
1,504.00
Save 163.90
Each
Ensures dependable performance for precise liquid handling. It is easy to install and supports consistent operation across a variety of laboratory applications.
  • Provides consistent performance for small-volume dispensing
  • Designed for compatibility with SMART+ systems
  • Helps ensure reproducibility across experiments
  • Reliable liquid handling for routine and sensitive applications
  • User-friendly design for quick setup and replacement
  • Supports efficient laboratory workflows
Specifications
AutoclavableAutoclavable
DescriptionSMART+ Small Metal Tip
For Use With (Application)Automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA.
For Use With (Equipment)Multidrop Combi SMART+
Quantity1 Ea.
Volume (Metric) Dead<1 mL
MaterialIn contact with liquid: PEEK, silicone, ruby, and stainless steel
Product TypeTube Dispensing Cassette
Volume (Metric)0.5 to 50 μL
Unit SizeEach

Frequently asked questions (FAQs)

What are the tubing lengths of the regular length and extended length cassettes for the Multidrop Combi Reagent Dispensers?

Regular length tubing (small and standard) cassette length is 40 cm long.

Extended length tubing (small and standard) cassette length is 200 cm long.
Note: You can cut the extended length tubing to the desired length but we recommend checking the calibration on the cassette, once modified, to ensure it is providing accurate results.