Multidrop™ Combi SMART Reagent Dispenser Accessories
Thermo Scientific™

Multidrop™ Combi SMART Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Quantity
N22704SMART+ Standard5 to 2500 μL1 Ea.
24073292SMART Small Plastic Tip0.5 to 50 μL1 Ea.
24073001SMART Small Plastic Tip0.5 to 50 μL5 Pk.
24072678SMART Long Standard5 to 2500 μL1 Ea.
24072676SMART Standard5 to 2500 μL5 Pk.
N22706SMART+ Long Standard5 to 2500 μL1 Ea.
Catalog number N22704
Price (USD)
1,281.60
Online exclusive
1,438.00
Save 156.40
Each
Description:
SMART+ Standard
Volume (Metric):
5 to 2500 μL
Quantity:
1 Ea.
Price (USD)
1,281.60
Online exclusive
1,438.00
Save 156.40
Each

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.

  • Designed for use with Multidrop Combi SMART+ dispensers
  • Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time
  • Available in standard and small tube versions
  • Distinguished from normal cassettes by antenna tags
  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
AutoclavableAutoclavable
DescriptionSMART+ Standard
For Use With (Application)Automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA.
For Use With (Equipment)Multidrop Combi SMART+ instrument with serial numbers 836-8C0001SM onwards
Quantity1 Ea.
Volume (Metric) Dead<7 mL
MaterialIn contact with liquid: PEEK, silicone, and PP (polypropylene)
Product TypeTube Dispensing Cassette
Volume (Metric)5 to 2500 μL
Unit SizeEach

Frequently asked questions (FAQs)

What are the tubing lengths of the regular length and extended length cassettes for the Multidrop Combi Reagent Dispensers?

Regular length tubing (small and standard) cassette length is 40 cm long.

Extended length tubing (small and standard) cassette length is 200 cm long.
Note: You can cut the extended length tubing to the desired length but we recommend checking the calibration on the cassette, once modified, to ensure it is providing accurate results.