Gibco™

Rat Fetal Neural Stem Cell Kit

Catalog number: N7744200
Gibco™

Rat Fetal Neural Stem Cell Kit

Catalog number: N7744200
Catalog Number
N7744200
also known as N7744-200
Unit Size
1 kit
Price (USD)
Price: 1,176.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
N7744200
also known as N7744-200
1 kit
Price: 1,176.00
Your Price:
-

Recurring order eligible

Save time by having your items shipped automatically.

Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Rat Fetal Neural Stem Cells are isolated and cryopreserved from the cortex of Sprague Dawley Rats at day E14 of gestation. A flexible, ready-to-use, and quality alternative to fresh cells, each vial contains 2 million cells per mL. Upon proliferation in StemPro NSC SFM, >75% of the cells retain their undifferentiated phenotype with the capability to differentiate into neurons, astrocytes, and oligodendrocytes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Neural Stem Cells
Classification
Serum-free
Culture Type
Mammalian Cell Culture
Form
Cryopreserved
Product Type
Neural Stem Cell Kit
Serum Level
Serum-free
Product Line
KnockOut™, StemPro™

Contents & Storage

• Rat Fetal Neural Stem Cells: store in liquid nitrogen
• KnockOut DMEM⁄F-12 Basal Medium: store at 2–8°C, protect from light
• StemPro NSC SFM Supplement: store at -5 to -20°C, protect from light
• FGF Basic Recombinant Human and EGF Recombinant Human: store at 2–8°C, desiccated