MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available inRead more
Have Questions?
Change view
Catalog Number
Quantity
N8010535
300 Strips
N8011535
1500 Strips (5 pk./300)
2 Options
Catalog number N8010535
Price (KRW)
168,000
Online offer
Ends: 31-Dec-2026
197,000
Save 29,000
Each
Quantity:
300 Strips
Price (KRW)
168,000
Online offer
Ends: 31-Dec-2026
197,000
Save 29,000
Each
MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. For real-time PCR applications, please see our MicroAmp™ Optical 8-Cap Strips (cat. no. 4323032), which have a flat-top profile. Attach the caps using the MicroAmp™ Cap Installing Tool.
Features:
• Will not interfere with sample reading • Provide tight seal to minimize evaporation • Are easy to apply and remove
Minimize Evaporation Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.
Easy to Use Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.
Colored for Organization These caps are available in different colors to help you keep track of your tubes during assay preparation.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers
Format300 Strip Caps
No. of Wells96
Product LineMicroAmp™
Quantity300 Strips
Shipping ConditionRoom Temperature
Closure TypeDomed Cap
ColorNatural
MaterialPolypropylene
Product Type8-Strip Cap
Unit SizeEach
Contents & Storage
Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color. Store at room temperature.
Frequently asked questions (FAQs)
Can I use caps on the 96-well plates on the QuantStudio 12K Flex system?
Yes. We recommend that you use Optical Flat caps, such as Cat. No. N8010535. Use of domed caps may damage the heated cover.