MicroAmp™ 96-Well Tray
Applied Biosystems™
MicroAmp™ 96-Well Tray
The Applied Biosystems™ MicroAmp™ 96-Well Tray, specifically designed to snugly hold up to 96 MicroAmp™ Reaction Tubes with Caps, is자세히 알아보기
Have Questions?
카탈로그 번호수량
N801054110 Trays
카탈로그 번호 N8010541
제품 가격(KRW)
105,000
온라인 행사
Ends: 31-Dec-2026
119,000
할인액 14,000 (12%)
Each
수량:
10 Trays
제품 가격(KRW)
105,000
온라인 행사
Ends: 31-Dec-2026
119,000
할인액 14,000 (12%)
Each
The Applied Biosystems™ MicroAmp™ 96-Well Tray, specifically designed to snugly hold up to 96 MicroAmp™ Reaction Tubes with Caps, is required for more efficient sample processing on Applied Biosystems™ 96-well thermal cyclers.
• Engineered for efficient sample processing
• Designed specifically for MicroAmp™ Reaction Tubes with Caps
• Manufactured to high specification, ensuring a quality product
Convenience and Performance
The MicroAmp™ 96-Well Tray allows you to organize your tubes while adding sample and mix. This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the thermal cycler. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for thermal cycling.
Note:
See user's manual or package insert for limited label license and trademark information.
사양
인증/적합성Certified DNA/RNase Free
용도(애플리케이션)PCR
용도(장비)GeneAmp 9700, 2720 Thermal Cycler
플레이트 호환성96-Well Plates
제품 유형Tray
수량10 Trays
배송 조건Room Temperature
색상Natural
제품라인MicroAmp
Unit SizeEach
구성 및 보관
Contains 10 MicroAmp™ 96-Well Trays. Store at room temperature.