The Applied Biosystems™ MicroAmp™ 96-Well Full Plate Cover enables you to seal up to 96 wells in MicroAmp™ Tray/Retainer SetRead more
Have Questions?
Catalog Number
Quantity
N8010550
5 covers
Catalog number N8010550
Price (KRW)
176,000
Online offer
Ends: 31-Dec-2026
207,000
Save 31,000 (15%)
Each
Quantity:
5 covers
Price (KRW)
176,000
Online offer
Ends: 31-Dec-2026
207,000
Save 31,000 (15%)
Each
The Applied Biosystems™ MicroAmp™ 96-Well Full Plate Cover enables you to seal up to 96 wells in MicroAmp™ Tray/Retainer Set or Reaction Plates to prevent evaporation during thermal cycling of precious samples. The durable design will allow for continuous use with high-quality results.
• Convenient, cost-saving alternative to MicroAmp™ Caps • Patented one-piece design for convenience • Guaranteed reusable up to five times
Note: See user's manual or package insert for limited label license, and trademark information.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Application)PCR
For Use With (Equipment)2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
Packaging5 Covers
Plate Compatibility96-Well Plates
Product TypeMicroplate Lid
Quantity5 covers
Shipping ConditionRoom Temperature
ColorNatural
No. of Wells96
Product LineMicroAmp
SterilityNon-sterile
Unit SizeEach
Contents & Storage
MicroAmp™ 96-Well Full Plate Cover is manufactured from PCR-compatible rubber. It is non-sterile, autoclavable and guaranteed reusable up to five times.