GeneAmp® PCR System 9700 Dual 384 well - FAQs
GeneAmp® PCR System 9700 Dual 384 well - FAQs
View additional product information for GeneAmp® PCR System 9700 Dual 384 well - FAQs (N8050002)
8 product FAQs found
How do I replace lost calibration files on the GeneAmp PCR System 9700?
A guide to replacing lost calibration files for the GeneAmp PCR System 9700 can be found here: http://tools.thermofisher.com/content/sfs/brochures/cms_042718.pdf.
How do I perform the temperature verification on the GeneAmp PCR System 9700?
Please refer to the guidelines in this protocol(https://tools.thermofisher.com/content/sfs/manuals/cms_070449.pdf) specific to the GeneAmp PCR System 9700 and block type.
How do I program an infinite hold in the GeneAmp PCR System 9700?
To add an infinite hold, set the time for the hold to ‘99.59'. The screen should then default to the infinity symbol for your hold.
Is the GeneAmp PCR System 9700 capable of running a gradient?
No, the GeneAmp PCR System 9700 does not have a gradient feature. Our newer thermal cyclers including the ProFlex PCR System and Veriti Thermal Cycler have better-than-gradient VeriFlex Blocks.
What is the Auto Delta (AutoX) feature on the GeneAmp PCR System 9700?
The AutoDelta feature allows you to incrementally increase or decrease the temperature or hold time with each cycle for a given step within a cycling stage. One common application for this is touchdown PCR.
What is the max block ramp rate of the GeneAmp PCR System 9700?
Please see the max block ramp rates in degrees/C below:
- 96-well aluminum: 2.3
- 96-well gold: 2.6
- 96-well silver: 2.6
- dual 96-well: 1.6
- dual 384-well: 1.8
- 60-well: 1.9
What types of plates or tubes can I use with the GeneAmp PCR System 9700?
The plates and plastics will be specific to the block type. Please refer to the tables here (https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics.html) for a list of compatible plastics, based on your block type.
What types of blocks are available on the GeneAmp PCR System 9700?
The GeneAmp PCR System 9700 offers interchangeable sample blocks in a variety of well formats and materials to accommodate a wide range of applications.
96-Well Aluminum
96-Well Gold
96-Well Silver
Dual 96-Well
Dual 384-Well
60-Well