GeneAmp® EZ rTth RNA PCR Kit - FAQs
GeneAmp® EZ rTth RNA PCR Kit - FAQs
View additional product information for GeneAmp® EZ rTth RNA PCR Kit - FAQs (N8080179)
1 product FAQs found
Does rTth DNA polymerase (Cat. No. N808-0098) have RNase H activity?
No, rTth DNA polymerase does not have RNase H activity.